Merck
  • Luminescent properties of CdTe quantum dots synthesized using 3-mercaptopropionic acid reduction of tellurium dioxide directly.
Luminescent properties of CdTe quantum dots synthesized using 3-mercaptopropionic acid reduction of tellurium dioxide directly.
Nanoscale research letters (2013-05-31)
Mao Shen, Wenping Jia, Yujing You, Yan Hu, Fang Li, Shidong Tian, Jian Li, Yanxian Jin, Deman Han
ABSTRACT
A facile one-step synthesis of CdTe quantum dots (QDs) in aqueous solution by atmospheric microwave reactor has been developed using 3-mercaptopropionic acid reduction of TeO2 directly. The obtained CdTe QDs were characterized by ultraviolet-visible spectroscopy, fluorescent spectroscopy, X-ray powder diffraction, multifunctional imaging electron spectrometer (XPS), and high-resolution transmission electron microscopy. Green- to red-emitting CdTe QDs with a maximum photoluminescence quantum yield of 56.68% were obtained.
Sigma-Aldrich
Tellurium dioxide, ≥99%