MilliporeSigma
  • Chlorpromazine and dopamine: conformational similarities that correlate with the antischizophrenic activity of phenothiazine drugs.
Chlorpromazine and dopamine: conformational similarities that correlate with the antischizophrenic activity of phenothiazine drugs.
Proceedings of the National Academy of Sciences of the United States of America (1971-10-01)
A S Horn, S H Snyder
ABSTRACT
Phenothiazines and butyrophenones are known to alter dopamine (3,4-dihydroxyphenethylamine) metabolism in the brain in a fashion suggesting that they may block dopamine receptors. We observed, using Dreiding molecular models, that dopamine in its solid-state conformation is superimposable upon a portion of the known x-ray structure of chlorpromazine [2-chloro-10-(3-dimethylaminopropyl)-phenothiazine]. The ability of phenothiazine drugs to mimic the dopamine-like conformation correlates with their antischizophrenic efficacy. These structure-activity relationships explain the importance of a substituent in ring a, a three-carbon side chain bearing the amino group, and a hetero atom between rings a and c.
Sigma-Aldrich
Phenothiazine, purum, ≥98.0% (GC)