MilliporeSigma
  • Purification of a young age-related glycoprotein (Ugl-Y) from normal human urine.
Purification of a young age-related glycoprotein (Ugl-Y) from normal human urine.
Journal of biochemistry (1987-02-01)
R Iida, T Yasuda, K Kishi
ABSTRACT
Ugl-Y is a glycoprotein that is detected in normal urine samples from young men and women aged 0 to 17 years. It was purified by ammonium sulfate precipitation and various column chromatographies including affinity chromatography using anti-adult urine antibody coupled to Sepharose 4B. The homogeneity of the glycoprotein was confirmed by polyacrylamide gel electrophoresis, isoelectric focusing, column chromatography on Sephadex G-75, and the precipitation reaction with anti-Ugl-Y antibody. It was shown to have a molecular weight of 29,000 by gel filtration, and to contain 5.2% neutral sugars (mannose and galactose) and 4% hexosamine (glucosamine). Amino acid analysis of the glycoprotein indicated high contents of acidic and hydroxylic amino acids. Its origin is unknown.