MilliporeSigma
  • Volatile organic compound identification and characterization by PCA and mapping at a high-technology science park.
Volatile organic compound identification and characterization by PCA and mapping at a high-technology science park.
Environmental pollution (Barking, Essex : 1987) (2014-07-16)
Cheng-Hang Lan, Yu-Li Huang, Sheng-Huei Ho, Chiung-Yu Peng
ABSTRACT
High-technology industries have grown continuously in Taiwan and elsewhere in the world. Volatile organic compounds (VOCs) comprise the highest percentage of emissions in these industries. The objectives of this study were to identify VOC sources and to apportion their contributions by using a three-step approach. These included estimating concentration distributions, performing principal component analysis (PCA), and mapping concentration contours. The results showed that the dominant compound groups were aromatic and aliphatic compounds. The PCA resolved four emission sources: vehicular traffic, industrial solvents, waste water plants, and cleaning/degreasing agents. Spatial distributions showed that concentrations of vehicular traffic-related compounds (benzene and isooctane) were highest at the entrances to the science park, and strongly related to traffic volume, and that the emissions of industry-related compounds (xylene and ethylbenzene) were closest to the associated sources. This study provided an accurate, practical and efficient method of characterizing emission sources in an industrial complex.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 1 %, NMR tube size 5 mm × 7 in.
Supelco
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 1% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d ("100%", 99.96 atom % D)
Supelco
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 8 mm × 8 in.
Supelco
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Benzene, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Benzene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Benzene, JIS special grade, ≥99.5%
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%
Supelco
Ethylbenzene, analytical standard
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Ethylbenzene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzene, analytical standard
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Ethylbenzene, ReagentPlus®, 99%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%