Merck
  • ATRP synthesis of amphiphilic random, gradient, and block copolymers of 2-(dimethylamino)ethyl methacrylate and n-butyl methacrylate in aqueous media.
ATRP synthesis of amphiphilic random, gradient, and block copolymers of 2-(dimethylamino)ethyl methacrylate and n-butyl methacrylate in aqueous media.
Biomacromolecules (2003-09-10)
Sang Beom Lee, Alan J Russell, Krzysztof Matyjaszewski
ABSTRACT
Amphiphilic random, gradient, and block copolymers of 2-(dimethylamino)ethyl methacrylate (DMAEMA) and n-butyl methacrylate (BMA) were synthesized by atom transfer radical polymerization (ATRP) in water/2-propanol mixtures using a methoxy-poly(ethylene glycol) (MPEG) (M(n) = 2000) macroinitiator. Kinetic studies indicate that the copolymerization is well controlled with molecular weights increasing linearly with conversion. Copolymers with molecular weights up to M(n) = 34000 and low polydispersities (M(w)/M(n) = 1.11-1.47) were prepared. The reactivity ratios were calculated for the copolymerizations catalyzed by CuBr/bpy, (r(DMAEMA) = 1.07, r(BMA) = 1.24). The thermosensitivity and aggregation properties of the random, gradient, and block copolymers significantly depended on the architecture of the copolymers. The lower critical solution temperature of MPEG-b-PDMAEMA(84) was 38 degrees C (5 wt % in water).
Sigma-Aldrich
2-(Dimethylamino)ethyl methacrylate, contains 700-1000 ppm monomethyl ether hydroquinone as inhibitor, 98%