MilliporeSigma
A chemically defined medium for the growth of Cowdria ruminantium.
The Onderstepoort journal of veterinary research (2001-06-14)
E Zweygarth, A I Josemans
ABSTRACT
Chemically defined media, termed SFMC-23 and SFMC-36, were devised for the in vitro culture of Cowdria ruminantium, the causative agent of heartwater in domestic ruminants. Both media were based on Dulbecco's modified Eagle's medium nutrient mixture Ham F-12 (DME/F-12) containing various supplements. Medium SFMC-23 and SFMC-36 supported the long-term growth of the Welgevonden stock of C. ruminantium for a total of 55 and 28 passages, respectively, with regular passage intervals of 3 days. Using SFMC-23, split ratios varied from 5-10, depending on which host cell line was used. Other stocks of C. ruminantium (Sankat, Blaauwkrantz, Senegal) were successfully propagated for a test period of ten passages.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10000 units
In Stock
Details...
Diagnostics Manufacturers should use alternative grade product: SRE0018
$101.00
25000 units
In Stock
Details...
$203.00
50000 units
In Stock
Details...
$373.00
200000 units
In Stock
Details...
$1,120.00
Sigma-Aldrich
Nutrient Mixture F-12 Ham, Kaighn′s Modification, with L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10000 units
In Stock
Details...
Diagnostics Manufacturers should use alternative grade product: SRE0018
$101.00
25000 units
In Stock
Details...
$203.00
50000 units
In Stock
Details...
$373.00
200000 units
In Stock
Details...
$1,120.00