MilliporeSigma
  • The rebalanced pathway significantly enhances acetoin production by disruption of acetoin reductase gene and moderate-expression of a new water-forming NADH oxidase in Bacillus subtilis.
The rebalanced pathway significantly enhances acetoin production by disruption of acetoin reductase gene and moderate-expression of a new water-forming NADH oxidase in Bacillus subtilis.
Metabolic engineering (2014-02-15)
Xian Zhang, Rongzhen Zhang, Teng Bao, Zhiming Rao, Taowei Yang, Meijuan Xu, Zhenghong Xu, Huazhong Li, Shangtian Yang
ABSTRACT
Bacillus subtilis produces acetoin as a major extracellular product. However, the by-products of 2,3-butanediol, lactic acid and ethanol were accompanied in the NADH-dependent pathways. In this work, metabolic engineering strategies were proposed to redistribute the carbon flux to acetoin by manipulation the NADH levels. We first knocked out the acetoin reductase gene bdhA to block the main flux from acetoin to 2,3-butanediol. Then, among four putative candidates, we successfully screened an active water-forming NADH oxidase, YODC. Moderate-expression of YODC in the bdhA disrupted B. subtilis weakened the NADH-linked pathways to by-product pools of acetoin. Through these strategies, acetoin production was improved to 56.7g/l with an increase of 35.3%, while the production of 2,3-butanediol, lactic acid and ethanol were decreased by 92.3%, 70.1% and 75.0%, respectively, simultaneously the fermentation duration was decreased 1.7-fold. Acetoin productivity by B. subtilis was improved to 0.639g/(lh).
Sigma-Aldrich
Acetoin, primarily dimer, ≥95%, FG