MilliporeSigma
  • The Sulfolobus rod-shaped virus 2 encodes a prominent structural component of the unique virion release system in Archaea.
The Sulfolobus rod-shaped virus 2 encodes a prominent structural component of the unique virion release system in Archaea.
Virology (2010-05-22)
Tessa E F Quax, Mart Krupovic, Soizick Lucas, Patrick Forterre, David Prangishvili
ABSTRACT
Recently a unique mechanism of virion release was discovered in Archaea, different from lysis and egress systems of bacterial and eukaryotic viruses. It involves formation of pyramidal structures on the host cell surface that rupture the S-layer and by opening outwards, create apertures through which mature virions escape the cell. Here we present results of a protein analysis of Sulfolobus islandicus cells infected with the rudivirus SIRV2, which enable us to postulate SIRV2-encoded protein P98 as the major constituent of these exceptional cellular ultrastructures.
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, suitable for protein sequencing, salt-free, lyophilized powder