MilliporeSigma
  • Inhibition of Rous sarcoma virus replication and cell transformation by a specific oligodeoxynucleotide.
Inhibition of Rous sarcoma virus replication and cell transformation by a specific oligodeoxynucleotide.
Proceedings of the National Academy of Sciences of the United States of America (1978-01-01)
P C Zamecnik, M L Stephenson
PMID75545
ABSTRACT
The tridecamer d(A-A-T-G-G-T-A-A-A-A-T-G-G), which is complementary to 13 nucleotides of the 3'- and 5'-reiterated terminal sequences of Rous sarcoma virus 35S RNA, was added to chick embryo fibroblast tissue cultures infected with Rous sarcoma virus. Inhibition of virus production resulted. The inference emerges that the tridecamer and its counterpart with blocked 3'- and 5'-hydroxyl termini enter the chick fibroblast cells, hybridize with the terminal reiterated sequences at the 3' and 5' ends of the 35S RNA, and interfere with one or more steps involved in viral production and cell transformation. Likely sites of action are (i) the circularization step of the proviral DNA intermediate, and (ii) the initiation of translation, the latter being described in the following communication [Stephenson, M. L. & Zamecnik, P. C. (1978) Proc. Natl. Acad. Sci. USA 75, 285--288].
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose and L-glutamine, without sodium bicarbonate and phenol red, powder, suitable for cell culture