MilliporeSigma
  • Interactions with single-stranded and double-stranded DNA-binding factors and alternative promoter conformation upon transcriptional activation of the Htf9-a/RanBP1 and Htf9-c genes.
Interactions with single-stranded and double-stranded DNA-binding factors and alternative promoter conformation upon transcriptional activation of the Htf9-a/RanBP1 and Htf9-c genes.
The Journal of biological chemistry (1998-02-07)
G Di Matteo, M Salerno, G Guarguaglini, B Di Fiore, F Palitti, P Lavia
ABSTRACT
The murine Htf9-a/RanBP1 and Htf9-c genes are divergently transcribed from a shared TATA-less promoter. Transcription of both genes is initiated on complementary DNA strands and is controlled by cell cycle-dependent mechanisms. The bidirectional promoter harbors a genomic footprint flanking the major transcription start site of both genes. Transient promoter assays showed that the footprinted element is important for transcription of both genes. Protein-binding experiments and antibody assays indicated that members of the retinoid X receptor family interact with the double-stranded site. In addition, distinct factors interact with single DNA strands of the element. Double-stranded binding factors were highly expressed in liver cells, in which neither gene is transcribed, while single-stranded binding proteins were abundant in cycling cells, in which transcription of both genes is efficient. In vivo S1 analysis of the promoter depicted an S1-sensitive organization in cells in which transcription of both genes is active; S1 sensitivity was not detected in conditions of transcriptional repression. Thus, the same element is a target for either retinoid X receptor factors, or for single-stranded binding proteins, and form distinct complexes in different cellular conditions depending on the DNA conformation in the binding site.