MilliporeSigma
  • A stable, commercially available sulfenyl chloride for the activation of thioglycosides in conjunction with silver trifluoromethanesulfonate.
A stable, commercially available sulfenyl chloride for the activation of thioglycosides in conjunction with silver trifluoromethanesulfonate.
Carbohydrate research (2008-04-01)
David Crich, Feng Cai, Fan Yang
ABSTRACT
p-Nitrobenzenesulfenyl chloride is a stable commercially available sulfenyl chloride that, in conjunction with silver triflate, cleanly activates a wide range of thioglycosides for glycosylation at -78 degrees C in CH(2)Cl(2).
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)