Merck
  • Flow injection amperometric detection of OP nerve agents based on an organophosphorus-hydrolase biosensor detector.
Flow injection amperometric detection of OP nerve agents based on an organophosphorus-hydrolase biosensor detector.
Biosensors & bioelectronics (2002-12-18)
Joseph Wang, Robin Krause, Kirstin Block, Mustafa Musameh, Ashok Mulchandani, Michael J Schöning
ABSTRACT
A flow-injection system with an organophosphorus-hydrolase (OPH)-biosensor detector has been developed and characterized for the rapid detection of organophosphorus (OP) nerve agents. The enzyme was immobilized onto a thin-film gold detector through a cystamine-glutaraldehyde coupling. Factors influencing the performance were optimized. The resulting flow system offered a fast, sensitive, selective, and stable response. The peak current increased linearly with the concentration of paraoxon and methyl parathion over the 1-10 microM range (sensitivity, 2.29 and 1.04 nA/microM, respectively). The OPH-biosensor flow injection systems offered low detection limits (e.g. 0.1 microM paraoxon), along with a good precision (R.S.D. of 3.6% for 20 successive injections of a 1.0 microM paraoxon solution). The OPH-biosensor flow detector offers great promise for rapid field screening of OP pesticides and nerve agents.
Sigma-Aldrich
Silicon dioxide, acid washed and calcined, Analytical Reagent