MilliporeSigma
  • Anti-inflammation effects of naloxone involve phosphoinositide 3-kinase delta and gamma.
Anti-inflammation effects of naloxone involve phosphoinositide 3-kinase delta and gamma.
The Journal of surgical research (2014-07-14)
Tao-Yeuan Wang, Nuan-Yen Su, Ping-Cheng Shih, Pei-Shan Tsai, Chun-Jen Huang
ABSTRACT
Phosphoinositide 3-kinase (PI3K) delta and gamma (the p110δ and p110γ isoforms of PI3K) actively participate in the process of inflammation. We sought to elucidate the possible roles of PI3Kδ and PI3Kγ in mediating the anti-inflammation effects of naloxone. Murine macrophages were treated with endotoxin, endotoxin plus naloxone, or endotoxin plus naloxone plus the PI3K inhibitors (the PI3Kδ inhibitor IC87114, the PI3Kγ inhibitor AS252424, or IC87114 plus AS252424) and denoted as the LPS, LPS + N, LPS + N + IC, LPS + N + AS, and LPS + N + IC + AS group, respectively. Differences in inflammatory molecules and levels of nuclear factor-κB (NF-κB) activation and Akt activation (indicator of PI3K activity) among these groups were compared. The concentrations of inflammatory molecules (macrophage inflammatory protein 2, tumor necrosis factor-α, interleukin-1β, and cyclooxygenase-2/prostaglandin E2) and the levels of NF-κB activation (p-NF-κB p65 and p-inhibitor-κB concentrations and NF-κB-DNA binding activity) of the LPS + N group were significantly lower than those of the LPS group (all P < 0.001). These data confirmed the anti-inflammation effects of naloxone. Moreover, the anti-inflammation effects of naloxone could be counteracted by the inhibitors of PI3Kδ and PI3Kγ, as the concentrations of inflammatory molecules and the levels of NF-κB activation of the LPS + N group were significantly lower than those of the LPS + N + IC, LPS + N + AS, and LPS + N + IC + AS groups (all P < 0.05). In contrast, the concentration of phosphorylated Akt of the LPS + N group was significantly higher than those of the LPS, LPS + N + IC, LPS + N + AS, and LPS + N + IC + AS groups (all P < 0.05). PI3Kδ and PI3Kγ play crucial roles in mediating the anti-inflammation effects of naloxone.
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Naloxone hydrochloride dihydrate, ≥98% (TLC and titration), powder
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Supelco
Naloxone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
Naloxone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Naloxone, United States Pharmacopeia (USP) Reference Standard