MilliporeSigma
  • Inverse associations of total and decaffeinated coffee with liver enzyme levels in National Health and Nutrition Examination Survey 1999-2010.
Inverse associations of total and decaffeinated coffee with liver enzyme levels in National Health and Nutrition Examination Survey 1999-2010.
Hepatology (Baltimore, Md.) (2014-08-16)
Qian Xiao, Rashmi Sinha, Barry I Graubard, Neal D Freedman
ABSTRACT
Coffee may have hepatoprotective effects and higher coffee consumption has been associated inversely with levels of liver enzymatic markers. However, it is unclear whether decaffeinated coffee is also associated with liver enzymes. The study population included 27,793 participants, age 20 or older, in the U.S. National Health and Nutrition Examination Survey (1999-2010). Coffee intake was evaluated by 24-hour dietary recall. Serum levels of alanine aminotransferase (ALT), aspartate aminotransferase (AST), alkaline phosphatase (ALP), and gamma-glutamyl transaminase (GGT) were measured. We examined the relationship between coffee intake and enzymatic levels using weighted multiple variable logistic (abnormally elevated levels of enzymes) and linear regression (continuous enzymatic levels). Total coffee consumption was inversely associated with abnormal levels of all four liver enzymes and continuous levels of AST, ALP, and GGT. Compared to those reporting no coffee consumption, participants reporting ≥ 3 cups per day had an odds ratio (OR; 95% confidence interval [CI]) of 0.75 (0.63, 0.89), 0.82 (0.68, 0.98), 0.73 (0.55, 0.95), and 0.69 (0.57, 0.83) for abnormal levels of ALT, AST, ALP, and GGT, respectively. Similar inverse associations were found with decaffeinated coffee intake and abnormal levels of ALT (OR (≥ 2 vs 0 cup/d): 0.62 [0.41, 0.94]), AST (0.74 [0.49, 1.11]), and GGT (0.70 [0.49-1.00]). Higher intakes of coffee, regardless of its caffeine content, were associated with lower levels of liver enzymes.
Product Description
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
γ-Glutamyltranspeptidase from porcine kidney, Type IV, 2.0-6.0 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
γ-Glutamyltranspeptidase from equine kidney, Type VI, 5-12 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
SKU
Pack Size
Availability
Price
Quantity
Supelco
Melting point standard 235-237°C, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
SKU
Pack Size
Availability
Price
Quantity
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Caffeine, BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥2,000 DEA units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
SKU
Pack Size
Availability
Price
Quantity
Caffeine, European Pharmacopoeia (EP) Reference Standard