MilliporeSigma
  • The intron-22-inverted F8 locus permits factor VIII synthesis: explanation for low inhibitor risk and a role for pharmacogenomics.
The intron-22-inverted F8 locus permits factor VIII synthesis: explanation for low inhibitor risk and a role for pharmacogenomics.
Blood (2014-11-20)
Zuben E Sauna, Jay N Lozier, Carol K Kasper, Chen Yanover, Timothy Nichols, Tom E Howard
ABSTRACT
Intron-22-inversion patients express the entire Factor VIII (FVIII)-amino-acid sequence intracellularly as 2 non-secreted polypeptides and have a positive "intracellular (I)-FVIII-CRM" status. Mutations conferring a positive I-FVIII-CRM status are associated with low inhibitor risk and are pharmacogenetically relevant because inhibitor risk may be affected by the nature of the therapeutic FVIII-protein (tFVIII), the affinity of any tFVIII-derived foreign peptide (tFVIII-fp) for any HLA class-II isomer (HLA-II) comprising individual major histocompatibility complex (MHC) repertoires, and the stability of any tFVIII-fp/HLA-II complex. We hypothesize that mutations conferring a completely or substantially negative I-FVIII-CRM status are pharmacogenetically irrelevant because inhibitor risk is high with any tFVIII and individual MHC repertoire.
Product Description
Human coagulation factor VIII concentrate, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
In Stock
Details...
$77.70
1 kg
In Stock
Details...
$136.00