MilliporeSigma
  • Multicolor live-cell chemical imaging by isotopically edited alkyne vibrational palette.
Multicolor live-cell chemical imaging by isotopically edited alkyne vibrational palette.
Journal of the American Chemical Society (2014-05-23)
Zhixing Chen, Daniel W Paley, Lu Wei, Andrew L Weisman, Richard A Friesner, Colin Nuckolls, Wei Min
ABSTRACT
Vibrational imaging such as Raman microscopy is a powerful technique for visualizing a variety of molecules in live cells and tissues with chemical contrast. Going beyond the conventional label-free modality, recent advance of coupling alkyne vibrational tags with stimulated Raman scattering microscopy paves the way for imaging a wide spectrum of alkyne-labeled small biomolecules with superb sensitivity, specificity, resolution, biocompatibility, and minimal perturbation. Unfortunately, the currently available alkyne tag only processes a single vibrational "color", which prohibits multiplex chemical imaging of small molecules in a way that is being routinely practiced in fluorescence microscopy. Herein we develop a three-color vibrational palette of alkyne tags using a (13)C-based isotopic editing strategy. We first synthesized (13)C isotopologues of EdU, a DNA metabolic reporter, by using the newly developed alkyne cross-metathesis reaction. Consistent with theoretical predictions, the mono-(13)C ((13)C≡(12)C) and bis-(13)C ((13)C≡(13)C) labeled alkyne isotopologues display Raman peaks that are red-shifted and spectrally resolved from the originally unlabeled ((12)C≡(12)C) alkynyl probe. We further demonstrated three-color chemical imaging of nascent DNA, RNA, and newly uptaken fatty-acid in live mammalian cells with a simultaneous treatment of three different isotopically edited alkynyl metabolic reporters. The alkyne vibrational palette presented here thus opens up multicolor imaging of small biomolecules, enlightening a new dimension of chemical imaging.
Product Description
Grace Bio-Labs SecureSeal imaging spacer, 8 wells, diam. × thickness 9 mm × 0.12 mm