Merck
  • Protracted effects of juvenile stressor exposure are mitigated by access to palatable food.
Protracted effects of juvenile stressor exposure are mitigated by access to palatable food.
PloS one (2014-05-08)
Jennifer Christine MacKay, Jonathan Stewart James, Christian Cayer, Pamela Kent, Hymie Anisman, Zul Merali
ABSTRACT
Stressor experiences during the juvenile period may increase vulnerability to anxiety and depressive-like symptoms in adulthood. Stressors may also promote palatable feeding, possibly reflecting a form of self-medication. The current study investigated the short- and long-term consequences of a stressor applied during the juvenile period on anxiety- and depressive-like behavior measured by the elevated plus maze (EPM), social interaction and forced swim test (FST). Furthermore, the effects of stress on caloric intake, preference for a palatable food and indices of metabolic syndrome and obesity were assessed. Male Wistar rats exposed to 3 consecutive days of variable stressors on postnatal days (PD) 27-29, displayed elevated anxiety-like behaviors as adults, which could be attenuated by consumption of a palatable high-fat diet. However, consumption of a palatable food in response to a stressor appeared to contribute to increased adiposity.
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Without calcium chloride, powder, suitable for cell culture