MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Pd-catalyzed Suzuki-Miyaura cross-coupling reactions between sulfamates and potassium Boc-protected aminomethyltrifluoroborates.
Pd-catalyzed Suzuki-Miyaura cross-coupling reactions between sulfamates and potassium Boc-protected aminomethyltrifluoroborates.
Organic letters (2013-05-08)
Gary A Molander, Inji Shin
ABSTRACT
Sulfamates were studied as the electrophilic partners in the palladium-catalyzed Suzuki-Miyaura cross-coupling reaction with potassium Boc-protected primary and secondary aminomethyltrifluoroborates. A broad range of substrates was successfully coupled to provide the desired products. Complex molecules containing a new carbon-carbon bond and an aminomethyl moiety could be prepared through this developed method.
Sigma-Aldrich
XPhos Pd G2, 98%