MilliporeSigma
  • Regulatory domains of the A-Myb transcription factor and its interaction with the CBP/p300 adaptor molecules.
Regulatory domains of the A-Myb transcription factor and its interaction with the CBP/p300 adaptor molecules.
The Biochemical journal (1997-06-15)
V Facchinetti, L Loffarelli, S Schreek, M Oelgeschläger, B Lüscher, M Introna, J Golay
ABSTRACT
The A-Myb transcription factor belongs to the Myb family of oncoproteins and is likely to be involved in the regulation of proliferation and/or differentiation of normal B cells and Burkitt's lymphoma cells. To characterize in detail the domains of A-Myb that regulate its function, we have generated a series of deletion mutants and have investigated their trans-activation potential as well as their DNA-binding activity. Our results have allowed us to delineate the trans-activation domain as well as two separate regulatory regions. The boundaries of the trans-activation domain (amino acid residues 218-319) are centred on a sequence rich in charged amino acids (residues 259-281). A region (residues 320-482) localized immediately downstream of the trans-activation domain and containing a newly identified conserved stretch of 48 residues markedly inhibits specific DNA binding. Finally the last 110 residues of A-Myb (residues 643-752), which include a sequence conserved in all mammalian myb genes (region III), negatively regulate the maximal trans-activation potential of A-Myb. We have also investigated the functional interaction between A-Myb and the nuclear adaptor molecule CBP [cAMP response element-binding protein (CREB)-binding protein]. We demonstrate that CBP synergizes with A-Myb in a dose-dependent fashion, and that this co-operative effect can be inhibited by E1A and can also be observed with the CBP homologue p300. We show that this functional synergism requires the presence of the A-Myb charged sequence and that it involves physical interaction between A-Myb and the CREB-binding domain of CBP.