MilliporeSigma
  • Peroxiredoxin 6, a 1-Cys peroxiredoxin, functions in antioxidant defense and lung phospholipid metabolism.
Peroxiredoxin 6, a 1-Cys peroxiredoxin, functions in antioxidant defense and lung phospholipid metabolism.
Free radical biology & medicine (2005-05-14)
Yefim Manevich, Aron B Fisher
ABSTRACT
Peroxiredoxin 6 (Prdx6), a bifunctional 25-kDa protein with both GSH peroxidase and phospholipase A2 activities, is the only mammalian 1-Cys member of the peroxiredoxin superfamily and is expressed in all major organs, with a particularly high level in lung. Prdx6 uses GSH as an electron donor to reduce H2O2 and other hydroperoxides including phospholipid hydroperoxides at approximately 5 micromol/mg protein/min with K1 approximately 3 x 10(6) M(-1) s(-1). Oxidation of the Cys47 to a sulfenic acid during catalysis requires piGST-catalyzed glutathionylation and reduction with GSH to complete the enzymatic cycle. Prdx6 stably overexpressed in cells protected against oxidative stress, whereas antisense treatment resulted in oxidant stress and apoptosis. Adenoviral-mediated overexpression of Prdx6 in mouse lungs protected against the toxicity of hyperoxia, whereas Prdx6-null mice were more sensitive to the effects of hyperoxia or paraquat. We postulate that Prdx6 functions in antioxidant defense mainly by facilitating repair of damaged cell membranes via reduction of peroxidized phospholipids. The PLA2 activity of Prdx6 is Ca2+ independent and maximal at acidic pH. Inhibition of PLA2 activity results in alterations of lung surfactant phospholipid synthesis and turnover. Thus, Prdx6, a unique mammalian peroxiredoxin, is an important antioxidant enzyme and has a major role in lung phospholipid metabolism.