MilliporeSigma
  • Simple and rapid determination of hydrogen peroxide using phosphine-based fluorescent reagents with sodium tungstate dihydrate.
Simple and rapid determination of hydrogen peroxide using phosphine-based fluorescent reagents with sodium tungstate dihydrate.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2006-06-15)
Maki Onoda, Takefumi Uchiyama, Ken-Ichi Mawatari, Kiyoko Kaneko, Kazuya Nakagomi
ABSTRACT
A simple batch method for the fluorometric determination of hydrogen peroxide using phosphine-based fluorescent reagents has been developed. A rapid, mild and selective derivatization reaction was achieved by adding sodium tungstate dihydrate to the reaction mixture of hydrogen peroxide and a phosphine-based fluorescent reagent. When 4-diphenylphosphino-7-methylthio-2,1,3-benzoxadiazole was used as a reagent, the derivatization reaction was completed after 2 min at room temperature. The calibration curve was linear between 12.5 and 500 ng hydrogen peroxide in a 10 microL sample solution. This method is accurate and has potential for on-line applications.
Sigma-Aldrich
Sodium tungstate dihydrate, ACS reagent, ≥99%