MilliporeSigma
  • Inhibitors of tick-borne flavivirus reproduction from structure-based virtual screening.
Inhibitors of tick-borne flavivirus reproduction from structure-based virtual screening.
ACS medicinal chemistry letters (2014-06-06)
Dmitry I Osolodkin, Liubov I Kozlovskaya, Evgenia V Dueva, Victor V Dotsenko, Yulia V Rogova, Konstantin A Frolov, Sergey G Krivokolysko, Ekaterina G Romanova, Alexey S Morozov, Galina G Karganova, Vladimir A Palyulin, Vladimir M Pentkovski, Nikolay S Zefirov
ABSTRACT
Flaviviruses form a large family of enveloped viruses affecting millions of people over the world. To date, no specific therapy was suggested for the infected people, making the treatment exclusively symptomatic. Several attempts were performed earlier for the design of fusion inhibitors for mosquito-borne flaviviruses, whereas for the tick-borne flaviviruses such design had not been performed. We have constructed homology models of envelope glycoproteins of tick-transmitted flaviviruses with the detergent binding pocket in the open state. Molecular docking of substituted 1,4-dihydropyridines and pyrido[2,1-b][1,3,5]thiadiazines was made against these models, and 89 hits were selected for the in vitro experimental evaluation. Seventeen compounds showed significant inhibition against tick-borne encephalitis virus, Powassan virus, or Omsk hemorrhagic fever virus in the 50% plaque reduction test in PEK cells. These compounds identified through rational design are the first ones possessing reproduction inhibition activity against tick-borne flaviviruses.
Sigma-Aldrich
Valerophenone, 99%