MilliporeSigma
  • ZnT-1 extrudes zinc from mammalian cells functioning as a Zn(2+)/H(+) exchanger.
ZnT-1 extrudes zinc from mammalian cells functioning as a Zn(2+)/H(+) exchanger.
Metallomics : integrated biometal science (2014-06-22)
Eden Shusterman, Ofer Beharier, Levy Shiri, Raz Zarivach, Yoram Etzion, Craig R Campbell, Il-Ha Lee, Ken Okabayashi, Anuwat Dinudom, David I Cook, Amos Katz, Arie Moran
ABSTRACT
ZnT-1 is a Cation Diffusion Facilitator (CDF) family protein, and is present throughout the phylogenetic tree from bacteria to humans. Since its original cloning in 1995, ZnT-1 has been considered to be the major Zn(2+) extruding transporter, based on its ability to protect cells against zinc toxicity. However, experimental evidence for ZnT-1 induced Zn(2+) extrusion was not convincing. In the present study, based on the 3D crystal structure of the ZnT-1 homologue, YiiP, that predicts a homodimer that utilizes the H(+) electrochemical gradient to facilitate Zn(2+) efflux, we demonstrate ZnT-1 dependent Zn(2+) efflux from HEK 293T cells using FluoZin-3 and Fura 2 by single cell microscope based fluorescent imaging. ZnT-1 facilitates zinc efflux in a sodium-independent, pH-driven and calcium-sensitive manner. Moreover, substitution of two amino acids in the putative zinc binding domain of ZnT-1 led to nullification of Zn(2+) efflux and rendered the mutated protein incapable of protecting cells against Zn(2+) toxicity. Our results demonstrate that ZnT-1 extrudes zinc from mammalian cells by functioning as a Zn(2+)/H(+) exchanger.