MilliporeSigma
  • Efficient, scalable and economical preparation of tris(deuterium)- and 13C-labelled N-methyl-N-nitroso-p-toluenesulfonamide (Diazald®) and their conversion to labelled diazomethane.
Efficient, scalable and economical preparation of tris(deuterium)- and 13C-labelled N-methyl-N-nitroso-p-toluenesulfonamide (Diazald®) and their conversion to labelled diazomethane.
Journal of labelled compounds & radiopharmaceuticals (2014-10-17)
Samuel W J Shields, Jeffrey M Manthorpe
ABSTRACT
A method for the preparation of multi-gramme quantities of N-methyl-d3-N-nitroso-p-toluenesulfonamide (Diazald-d3) and N-methyl-(13)C-N-nitroso-p-toluenesulfonamide (Diazald-(13)C) and their conversion to diazomethane-d2 and diazomethane-(13) C, respectively, is presented. This approach uses robust and reliable chemistry, and critically, employs readily commercially available and inexpensive methanol as the label source. Several reactions of labelled diazomethane are also reported, including alkene cyclopropanation, phenol methylation and α-diazoketone formation, as well as deuterium scrambling in the preparation of diazomethane-d2 and subsequent methyl esterification of benzoic acid.
Sigma-Aldrich
Sodium methoxide, reagent grade, 95%, powder
Sigma-Aldrich
Sodium methoxide solution, 25 wt. % in methanol
Sigma-Aldrich
Sodium methoxide solution, ACS reagent, 0.5 M CH3ONa in methanol (0.5N)