MilliporeSigma
  • Highly selective and sensitive paper-based colorimetric sensor using thiosulfate catalytic etching of silver nanoplates for trace determination of copper ions.
Highly selective and sensitive paper-based colorimetric sensor using thiosulfate catalytic etching of silver nanoplates for trace determination of copper ions.
Analytica chimica acta (2015-03-04)
Sudkate Chaiyo, Weena Siangproh, Amara Apilux, Orawon Chailapakul
ABSTRACT
A novel, highly selective and sensitive paper-based colorimetric sensor for trace determination of copper (Cu(2+)) ions was developed. The measurement is based on the catalytic etching of silver nanoplates (AgNPls) by thiosulfate (S2O3(2-)). Upon the addition of Cu(2+) to the ammonium buffer at pH 11, the absorption peak intensity of AuNPls/S2O3(2-) at 522 nm decreased and the pinkish violet AuNPls became clear in color as visible to the naked eye. This assay provides highly sensitive and selective detection of Cu(2+) over other metal ions (K(+), Cr(3+), Cd(2+), Zn(2+), As(3+), Mn(2+), Co(2+), Pb(2+), Al(3+), Ni(2+), Fe(3+), Mg(2+), Hg(2+) and Bi(3+)). A paper-based colorimetric sensor was then developed for the simple and rapid determination of Cu(2+) using the catalytic etching of AgNPls. Under optimized conditions, the modified AgNPls coated at the test zone of the devices immediately changes in color in the presence of Cu(2+). The limit of detection (LOD) was found to be 1.0 ng mL(-1) by visual detection. For semi-quantitative measurement with image processing, the method detected Cu(2+) in the range of 0.5-200 ng mL(-1)(R(2)=0.9974) with an LOD of 0.3 ng mL(-1). The proposed method was successfully applied to detect Cu(2+) in the wide range of real samples including water, food, and blood. The results were in good agreement according to a paired t-test with results from inductively coupled plasma-optical emission spectrometry (ICP-OES).
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride, beads, 98%
Sigma-Aldrich
Manganese(II) chloride, powder and chunks, ≥99% trace metals basis
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium thiosulfate solution, 1 M
Sigma-Aldrich
Copper(II) sulfate, JIS first grade, ≥97.5%
Sigma-Aldrich
Potassium chloride solution, 0.01 M
Sigma-Aldrich
Sodium thiosulfate solution, 0.01 M
Sigma-Aldrich
Sodium thiosulfate solution, 0.1 M
Sigma-Aldrich
Sodium thiosulfate solution, 0.025 M
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium Thiosulfate Solution, 2 g/dL in deionized water
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)