MilliporeSigma
  • Glycyrrhizin conjugated chitosan nanoparticles for hepatocyte-targeted delivery of lamivudine.
Glycyrrhizin conjugated chitosan nanoparticles for hepatocyte-targeted delivery of lamivudine.
The Journal of pharmacy and pharmacology (2014-03-20)
Deepak Mishra, Nivrati Jain, Vaibhav Rajoriya, Ashish K Jain
ABSTRACT
The present study was focused to prepare controlled release glycyrrhizin (GL) conjugated low molecular weight chitosan nanoparticles (CS-NPs) for liver targeting. The hydrophilic antiretroviral drug lamivudine was chosen as a model drug and encapsulated within glycyrrhizin conjugated low molecular weight chitosan nanoparticles (GL-CS-NPs) for liver specificity. First, the low molecular weight chitosan (CS) was synthesized through depolymerization method. The low molecular weight chitosan nanoparticles were prepared by inotropic gelation method. Then glycyrrhizin was conjugated with previously prepared low molecular weight chitosan nanoparticles (CS-NPs) and conjugation was confirmed by Fourier transform infrared (FT-IR) spectroscopy. The prepared GL-CS-NPs were characterized using dynamic light scattering, transmission electron microscopy and FT-IR. The encapsulation efficiency and in-vitro drug release behaviour of drug-loaded GL-CS-NPs were studied using ultra violet spectroscopy and high performance liquid chromatographic methods. Release of lamivudine from the nanoparticles exhibited a biphasic pattern, initial burst release and consequently sustained release. In-vivo biodistribution study suggested the target ability of GL-CS-NPs is better and haematological study shows decline of the tissue damage in comparison with plain drug solution. The experimental results show that the glycyrrhizin conjugated LMWC nanoparticles may be used as a potential drug delivery system with hepatocyte-targeting characteristics.
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice), ≥95.0% (NT)
Supelco
Glycyrrhizic acid ammonium salt, analytical standard, suitable for HPLC
USP
Lamivudine, United States Pharmacopeia (USP) Reference Standard
Supelco
Lamivudine, Pharmaceutical Secondary Standard; Certified Reference Material
Lamivudine for system suitability 1, European Pharmacopoeia (EP) Reference Standard
Ammonium glycyrrhizate, European Pharmacopoeia (EP) Reference Standard
Lamivudine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium (meta)periodate, ≥99.0%
Sigma-Aldrich
Lamivudine, ≥98% (HPLC), powder
Sigma-Aldrich
Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice), ≥70% (HPLC)
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Glycyrrhizate monoammonium, European Pharmacopoeia (EP) Reference Standard
Lamivudine for system suitability 2, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium persulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium periodate, ACS reagent, ≥99.8%