MilliporeSigma
  • Combination of a peroxisome proliferator-activated receptor-gamma agonist and an angiotensin II receptor blocker attenuates myocardial fibrosis and dysfunction in type 2 diabetic rats.
Combination of a peroxisome proliferator-activated receptor-gamma agonist and an angiotensin II receptor blocker attenuates myocardial fibrosis and dysfunction in type 2 diabetic rats.
Journal of diabetes investigation (2014-11-21)
Chi Young Shim, Byeong-Wook Song, Min-Ji Cha, Ki-Chul Hwang, Sungha Park, Geu-Ru Hong, Seok-Min Kang, Jong Eun Lee, Jong-Won Ha, Namsik Chung
ABSTRACT
We aimed to examine the effect of an angiotensin II receptor blocker (ARB), a peroxisome proliferator-activated receptor (PPAR)-gamma agonist, and their combination on myocardial fibrosis and function in type 2 diabetic rats. Five male Long-Evans Tokushima Otsuka (LETO) rats and 20 male Otsuka Long-Evans Tokushima Fatty (OLETF) rats were used. OLETF rats were assigned to four groups (n = 5 per group) at 28 weeks-of-age: untreated, losartan-treated, rosiglitazone-treated and combination-treated. The ARB, losartan, was administered at a dose of 5 mg/kg/day, and the PPAR-gamma agonist, rosiglitazone, was administered at a dose of 3 mg/kg/day by oral gavage for 12 weeks. Urine and blood samples were collected, and two-dimensional echocardiograms and strain rate imaging were obtained at 28 and 40 weeks. Cytokines were evaluated by reverse transcriptase polymerase chain reaction, and histological analysis was carried out at 40 weeks. At 40 weeks, the global radial strains of the losartan-treated (55.7 ± 4.5%, P = 0.021) and combination-treated groups (59.3 ± 6.7%, P = 0.001) were significantly higher compared with the untreated OLETFs (44.3 ± 10.5%). No difference was observed when compared with LETO rats. Although the rosiglitazone-treated group showed a better metabolic profile than the untreated OLETF group, there was no difference in the global radial strain (49.8 ± 6.0 vs 44.3 ± 10.5, P = 0.402). The expression of pro-inflammatory cytokines, and collagen type I and III were consistently attenuated in the losartan-treated and combination-treated OLETF groups, but not in the rosiglitazone-treated group. A combination of rosiglitazone and losartan attenuates myocardial fibrosis and dysfunction in type 2 diabetic rats.
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, 10 mg/mL
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Ethidium bromide, ≥95.0%
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder