MilliporeSigma
  • Phosphorylation of myofibrillogenesis regulator-1 activates the MAPK signaling pathway and induces proliferation and migration in human breast cancer MCF7 cells.
Phosphorylation of myofibrillogenesis regulator-1 activates the MAPK signaling pathway and induces proliferation and migration in human breast cancer MCF7 cells.
FEBS letters (2014-07-30)
Yuyan Gong, Hongwei He, Hong Liu, Caixia Zhang, Wuli Zhao, Rong-Guang Shao
ABSTRACT
Myofibrillogenesis regulator-1 (MR-1) has been characterized as a tumor promoter in many cancers. However, its mechanism of action has not been fully elucidated. Here, we report that MR-1 is overexpressed in human breast cancer cells and participates in tumor promotion in human breast cancer MCF7 cells by activating the ERK1/2 signaling pathway. MR-1 interacts with MEK1/2 and ERK1, and its N-terminal sequence plays a major role in promoting the MEK/ERK cascade. Furthermore, six phosphorylation sites of MR-1 were identified, and phosphorylation at S46 was shown to be critical for the activation of MEK/ERK. Therefore, our findings suggest that MR-1 functions as a tumor promoter in MCF7 cells by activating the MEK/ERK signaling.