MilliporeSigma
  • Plasma levels of dimethylarginines in preterm very low birth weight neonates: its relation with perinatal factors and short-term outcome.
Plasma levels of dimethylarginines in preterm very low birth weight neonates: its relation with perinatal factors and short-term outcome.
International journal of molecular sciences (2014-12-30)
Rob M Moonen, Maurice J Huizing, Giacomo Cavallaro, Gema E González-Luis, Pilar Bas-Suárez, Jaap A Bakker, Eduardo Villamor
ABSTRACT
Endogenously produced inhibitors of nitric oxide (NO) synthase, in particular asymmetric dimethylarginine (ADMA), are currently considered of importance in various disease states characterized by reduced NO availability. We investigated the association between plasma levels of ADMA, symmetric dimethylarginine (SDMA), L-arginine, and citrulline and perinatal factors and outcome in 130 preterm (gestational age ≤ 30 weeks) very low birth weight (VLBW, < 1500 g) infants. Plasma samples were collected 6-12 h after birth. We did not find significant correlations between ADMA, SDMA, L-arginine, and citrulline levels and gestational age or birth weight. However, the arginine:ADMA ratio (AAR, a better indicator of NO availability than either arginine or ADMA separately) was positively correlated with gestational age. ADMA and arginine levels were not significantly different between males and females but males showed a negative correlation between ADMA levels and gestational age. Perinatal factors such as preeclampsia, chrorioamnionitis, prolonged rupture of membranes, or form of delivery did not significantly alter dimethylarginine levels or AAR. In contrast, the AAR was significantly reduced in the infants with respiratory distress, mechanical ventilation, and systemic hypotension Therefore, our data suggest that altered NO availability may play a role in the respiratory and cardiovascular adaptation in preterm VLBW infants.
Sigma-Aldrich
NG,NG-Dimethylarginine dihydrochloride
SKU
Pack Size
Availability
Price
Quantity
Arginine, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
NG,NG′-Dimethyl-L-arginine di(p-hydroxyazobenzene-p′-sulfonate) salt, ≥99% (TLC)