MilliporeSigma
  • Inhibitory effects of emodin, baicalin, schizandrin and berberine on hefA gene: treatment of Helicobacter pylori-induced multidrug resistance.
Inhibitory effects of emodin, baicalin, schizandrin and berberine on hefA gene: treatment of Helicobacter pylori-induced multidrug resistance.
World journal of gastroenterology (2015-04-22)
Yan-Qiang Huang, Gan-Rong Huang, Ming-Hui Wu, Hua-Ying Tang, Zan-Song Huang, Xi-Han Zhou, Wen-Qiang Yu, Jian-Wei Su, Xiao-Qiang Mo, Bing-Pu Chen, Li-Juan Zhao, Xiao-Feng Huang, Hong-Yu Wei, Lian-Deng Wei
ABSTRACT
To investigate the inhibitory effects of emodin, baicalin, etc. on the hefA gene of multidrug resistance (MDR) in Helicobacter pylori (H. pylori). The double dilution method was used to screen MDR H. pylori strains and determine the minimum inhibitory concentrations (MICs) of emodin, baicalin, schizandrin, berberine, clarithromycin, metronidazole, tetracycline, amoxicillin and levofloxacin against H. pylori strains. After the screened MDR stains were treated with emodin, baicalin, schizandrin or berberine at a 1/2 MIC concentration for 48 h, changes in MICs of amoxicillin, tetracycline, levofloxacin, metronidazole and clarithromycin were determined. MDR strains with reduced MICs of amoxicillin were selected to detect the hefA mRNA expression by real-time quantitative PCR. A total of four MDR H. pylori strains were screened. Treatment with emodin, baicalin, schizandrin and berberine significantly decreased the MICs of amoxicillin and tetracycline against some strains, decreased by 1 to 2 times, but did not significantly change the MICs of clarithromycin, levofloxacin, and metronidazole against MDR strains. In the majority of strains with reduced MICs of amoxicillin, hefA mRNA expression was decreased; one-way ANOVA (SPSS 12.0) used for comparative analysis, P < 0.05. Emodin, baicalin, schizandrin and berberine significantly decreased the MICs of amoxicillin and tetracycline against some H. pylori strains, possibly by mechanisms associated with decreasing hefA mRNA expression.
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Supelco
Clarithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Clarithromycin, European Pharmacopoeia (EP) Reference Standard
USP
Clarithromycin, United States Pharmacopeia (USP) Reference Standard
Clarithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
USP
Amoxicillin, United States Pharmacopeia (USP) Reference Standard
USP
Clarithromycin Identity, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis