MilliporeSigma
  • Complete genome sequence of the caprolactam-degrading bacterium Pseudomonas mosselii SJ10 isolated from wastewater of a nylon 6 production plant.
Complete genome sequence of the caprolactam-degrading bacterium Pseudomonas mosselii SJ10 isolated from wastewater of a nylon 6 production plant.
Journal of biotechnology (2014-12-03)
Gun-Seok Park, Ji-Hun Chu, Sung-Jun Hong, Yunyoung Kwak, Abdur Rahim Khan, Byung Kwon Jung, Ihsan Ullah, Jae-Ho Shin
ABSTRACT
Pseudomonas mosselii strain SJ10 is a caprolactam-degrading bacterium belonging to the class Gammaproteobacteria, which was isolated from wastewater of the nylon 6 producing Seongseo industrial complex in Daegu, Republic of Korea. Here, we report the complete genome sequence of the strain, providing genetic information for biodegradation of aromatic compounds.
Product Description
Supelco
ε-Caprolactam, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
ε-Caprolactam, 99%