MilliporeSigma
  • Identification of QTLs associated with oil content and mapping FAD2 genes and their relative contribution to oil quality in peanut (Arachis hypogaea L.).
Identification of QTLs associated with oil content and mapping FAD2 genes and their relative contribution to oil quality in peanut (Arachis hypogaea L.).
BMC genetics (2014-12-11)
Manish K Pandey, Ming Li Wang, Lixian Qiao, Suping Feng, Pawan Khera, Hui Wang, Brandon Tonnis, Noelle A Barkley, Jianping Wang, C Corley Holbrook, Albert K Culbreath, Rajeev K Varshney, Baozhu Guo
ABSTRACT
Peanut is one of the major source for human consumption worldwide and its seed contain approximately 50% oil. Improvement of oil content and quality traits (high oleic and low linoleic acid) in peanut could be accelerated by exploiting linked markers through molecular breeding. The objective of this study was to identify QTLs associated with oil content, and estimate relative contribution of FAD2 genes (ahFAD2A and ahFAD2B) to oil quality traits in two recombinant inbred line (RIL) populations. Improved genetic linkage maps were developed for S-population (SunOleic 97R × NC94022) with 206 (1780.6 cM) and T-population (Tifrunner × GT-C20) with 378 (2487.4 cM) marker loci. A total of 6 and 9 QTLs controlling oil content were identified in the S- and T-population, respectively. The contribution of each QTL towards oil content variation ranged from 3.07 to 10.23% in the S-population and from 3.93 to 14.07% in the T-population. The mapping positions for ahFAD2A (A sub-genome) and ahFAD2B (B sub-genome) genes were assigned on a09 and b09 linkage groups. The ahFAD2B gene (26.54%, 25.59% and 41.02% PVE) had higher phenotypic effect on oleic acid (C18:1), linoleic acid (C18:2), and oleic/linoleic acid ratio (O/L ratio) than ahFAD2A gene (8.08%, 6.86% and 3.78% PVE). The FAD2 genes had no effect on oil content. This study identified a total of 78 main-effect QTLs (M-QTLs) with up to 42.33% phenotypic variation (PVE) and 10 epistatic QTLs (E-QTLs) up to 3.31% PVE for oil content and quality traits. A total of 78 main-effect QTLs (M-QTLs) and 10 E-QTLs have been detected for oil content and oil quality traits. One major QTL (more than 10% PVE) was identified in both the populations for oil content with source alleles from NC94022 and GT-C20 parental genotypes. FAD2 genes showed high effect for oleic acid (C18:1), linoleic acid (C18:2), and O/L ratio while no effect on total oil content. The information on phenotypic effect of FAD2 genes for oleic acid, linoleic acid and O/L ratio, and oil content will be applied in breeding selection.
Sigma-Aldrich
Oleic acid, SAJ first grade, ≥70.0%
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Supelco
Oleic acid, analytical standard
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Heptane, anhydrous, 99%
Supelco
Heptane, HPLC grade, ≥99%
Sigma-Aldrich
Heptane, SAJ first grade, ≥98.0%
Sigma-Aldrich
Heptane, JIS special grade, ≥99.0%
Supelco
Heptane, analytical standard
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Supelco
Heptane, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Heptane, ReagentPlus®, 99%
Sigma-Aldrich
Heptane, puriss. p.a., reag. Ph. Eur., ≥99% n-heptane basis (GC)
Sigma-Aldrich
Heptane, HPLC Plus, for HPLC, GC, and residue analysis, 99%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥96%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥99%