MilliporeSigma
A visual strip sensor for determination of iron.
Analytica chimica acta (2014-12-03)
Sanjukta A Kumar, Neha Thakur, Harshala J Parab, Shailaja P Pandey, Rakesh N Shinde, Ashok K Pandey, Sangita D Kumar, A V R Reddy
ABSTRACT
A visual strip has been developed for sensing iron in different aqueous samples like natural water and fruit juices. The sensor has been synthesized by UV-radiation induced graft polymerization of acrylamide monomer in microporous poly(propylene) base. For physical immobilization of iron selective reagent, the in situ polymerization of acrylamide has been carried out in the presence of 1,10-phenanthroline. The loaded strip on interaction with Fe(II) in aqueous solution turned into orange red color and the intensity of the color was found to be directly proportional to the amount of Fe(II) in the aqueous sample. The minimal sensor response with naked eye was found for 50ngmL(-1) of Fe in 15min of interaction. However, as low as 20ngmL(-1) Fe could be quantified using a spectrophotometer. The detection limit calculated using the 3s/S criteria, where 's' is the standard deviation of the absorbance of blank reagent loaded strip and 'S' is the slope of the linear calibration plot, was 1.0ngmL(-1). The strip was applied to measure Fe in a variety of samples such as ground water and fruit juices.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Acrylamide, SAJ first grade, ≥98.0%
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acrylamide, ≥99.9%
Sigma-Aldrich
Acrylamide, ≥98.0%
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Supelco
N,N-Dimethylformamide, analytical standard
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetophenone, ≥98%, FG
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend