Merck
  • Synaptic activity controls localization and function of CtBP1 via binding to Bassoon and Piccolo.
Synaptic activity controls localization and function of CtBP1 via binding to Bassoon and Piccolo.
The EMBO journal (2015-02-06)
Daniela Ivanova, Anika Dirks, Carolina Montenegro-Venegas, Cornelia Schöne, Wilko D Altrock, Claudia Marini, Renato Frischknecht, Denny Schanze, Martin Zenker, Eckart D Gundelfinger, Anna Fejtova
ABSTRACT
Persistent experience-driven adaptation of brain function is associated with alterations in gene expression patterns, resulting in structural and functional neuronal remodeling. How synaptic activity-in particular presynaptic performance-is coupled to gene expression in nucleus remains incompletely understood. Here, we report on a role of CtBP1, a transcriptional co-repressor enriched in presynapses and nuclei, in the activity-driven reconfiguration of gene expression in neurons. We demonstrate that presynaptic and nuclear pools of CtBP1 are interconnected and that both synaptic retention and shuttling of CtBP1 between cytoplasm and nucleus are co-regulated by neuronal activity. Finally, we show that CtBP1 is targeted and/or anchored to presynapses by direct interaction with the active zone scaffolding proteins Bassoon and Piccolo. This association is regulated by neuronal activity via modulation of cellular NAD/NADH levels and restrains the size of the CtBP1 pool available for nuclear import, thus contributing to the control of activity-dependent gene expression. Our combined results reveal a mechanism for coupling activity-induced molecular rearrangements in the presynapse with reconfiguration of neuronal gene expression.
Sigma-Aldrich
Abberior® STAR 580, NHS ester, for STED application