MilliporeSigma
  • Multiplexed electrochemical immunoassay using streptavidin/nanogold/carbon nanohorn as a signal tag to induce silver deposition.
Multiplexed electrochemical immunoassay using streptavidin/nanogold/carbon nanohorn as a signal tag to induce silver deposition.
Analytica chimica acta (2014-09-30)
Changrong Zhao, Jie Wu, Huangxian Ju, Feng Yan
ABSTRACT
An ultrasensitive multiplexed immunoassay method was developed by using streptavidin/nanogold/carbon nanohorn (SA/Au/CNH) as a novel signal tag to induce silver enhancement for signal amplification. The Au/CNH was prepared by in situ growth of nanogold on carboxylated CNH and functionalized with streptavidin. The SA/Au/CNH showed well dispersibility in physiological buffer and could sever as a common tracing tag to recognize biotinylated signal antibody. The immunosensor array was prepared on disposable screen-printed electrodes. Through sandwich-type immunoreaction and biotin-streptavidin affinity reaction, the SA/Au/CNH tag was captured on the immunoconjugates to induce silver deposition and amplify the electrochemical stripping signals. Using α-fetoprotein and carcinoembryonic antigen as model analytes, the proposed method showed wide linear ranges with the detection limits down to 0.024 pg mL(-1) and 0.032 pg mL(-1), respectively, and eliminated completely signal cross-talk between adjacent immunosensors. It provided a convenient, high-efficient and ultrasensitive electrochemical detection route for biological analytes, showing great potential in clinical application.
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
VenPure® SF, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Diallyldimethylammonium chloride, ≥97.0% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Diallyldimethylammonium chloride solution, 65 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%