MilliporeSigma
  • Two new alpha chain variants: Hb Boghé [alpha58(E7)His-->Gln, alpha2], a variant on the distal histidine, and Hb CHarolles [alpha103(G10)His-Tyr, alpha1].
Two new alpha chain variants: Hb Boghé [alpha58(E7)His-->Gln, alpha2], a variant on the distal histidine, and Hb CHarolles [alpha103(G10)His-Tyr, alpha1].
Hemoglobin (1999-11-24)
P Lacan, A Francina, G Souillet, M Aubry, N Couprie, L Dementhon, M Becchi
ABSTRACT
The present paper reports two new alpha-globin chain variants: Hb Boghé [alpha58(E7)His-->Gln, alpha2] and Hb Charolles [alpha103(G10)His-->Tyr, alpha1]. Hb Boghé was found in a 12-month-old girl who was treated for malignant histiocytosis at 9 months of age and received a bone marrow transplant from her sister. Hb Boghé was undetectable by isoelectrofocusing and high performance liquid chromatography of hemoglobins. It was only revealed by polyacrylamide gel electrophoresis of globin chains in the presence of urea-Triton X-100 and accounted for 10% of the total hemoglobin. Hb Charolles was detected in a 46-year-old patient who presented with microcytosis and hypochromia. It was easily detected by isoelectrofocusing and high performance liquid chromatography. Hb Charolles accounted for 11% of the total hemoglobin. Characterization of the two hemoglobin variants was achieved by DNA and restriction enzyme analyses. Oxygen equilibrium curves measured on whole blood with Hb Boghé were normal. DNA sequencing revealed the association of Hb Charolles with a common mutation of the alpha2 polyadenylation site: AATAAA-->AATAAG.