MilliporeSigma
  • 100 selective oxygenation of p-xylene to p-tolualdehyde via photoinduced electron transfer
100 selective oxygenation of p-xylene to p-tolualdehyde via photoinduced electron transfer
Organic letters (2000-11-14)
Ohkubo, Fukuzumi
ABSTRACT
The 100% selective oxygenation of p-xylene to p-tolualdehyde is initiated by photoinduced electron transfer from p-xylene to the singlet excited state of 10-methyl-9-phenylacridinium ion under visible light irradiation, yielding p-tolualdehyde exclusively as the final oxygenated product. The reason for the high selectivity in the photocatalytic oxygenation of p-xylene is discussed on the basis of the photoinduced electron transfer mechanism.