MilliporeSigma
Radiopacity of dual-cured and chemical-cured resin-based cements.
The International journal of prosthodontics (1998-05-20)
M H Rubo, O el-Mowafy
ABSTRACT
The objective of this study was to determine radiopacity values of a group of resin-based cements and to compare them to those of enamel and dentin. Three specimens 2.5 mm thick were prepared from each of 18 cements (nine chemical-cured and nine dual-cured). Three tooth sections 2.5 mm thick were cut from extracted molars. Specimens and tooth sections were divided into three matching groups. Following standard radiographic techniques, images of each group of specimens, along with an aluminum step wedge were obtained. Optical density readings for each material were determined with a transmission densitometer. Radiopacity values were subsequently calculated as equivalents of aluminum thickness. Analysis of variance indicated significant differences in radiopacity values among the materials (P < 0.0001). Four dual-cured cements (Variolink, Geristore, Enforce, and Nexus) had radiopacity values significantly greater than that of enamel, while two (Choice and Adherence) had radiopacity values similar to enamel. For the remaining three dual-cured cements, Duolink had a radiopacity value significantly lower than that of enamel but higher than that of dentin, while both Lute-It and Resinomer had values similar to dentin. For the chemical-cured cements, five materials had radiopacity values significantly higher than enamel (Sealbond, Advance, Scotchbond, Cement-It, and Dyract-Cem), while two had values significantly lower than enamel but higher than dentin (Biomer and Panavia 21). The remaining two cements had radiopacity values significantly lower than dentin. For a group of 18 resin-based cements, some had radiopacity values similar to or significantly lower than that of dentin.
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit