MilliporeSigma
  • cDNA cloning, expression and chromosome mapping of the human STAT4 gene: both STAT4 and STAT1 genes are mapped to 2q32.2-->q32.3.
cDNA cloning, expression and chromosome mapping of the human STAT4 gene: both STAT4 and STAT1 genes are mapped to 2q32.2-->q32.3.
Cytogenetics and cell genetics (1997-01-01)
K Yamamoto, H Kobayashi, A Arai, O Miura, S Hirosawa, N Miyasaka
ABSTRACT
Studies of transcriptional activation by interferons and a variety of cytokines have led to the identification of a family of proteins that serve as signal transducers and activators of transcription (STAT). STAT4 is phosphorylated following interleukin (IL)-12 stimulation and is essential for IL-12 signal transduction. The human STAT4 cDNA was cloned, and both STAT4 and STAT1 genes were mapped to human chromosome bands 2q32.2-->q32.3 by fluorescence in situ hybridization. These results suggest that STAT4 and STAT1 may have arisen via a tandem gene duplication. However, human STAT1 is expressed ubiquitously, whereas human STAT4 is expressed in several tissues including spleen, heart, brain, peripheral blood cells, and testis.
Sigma-Aldrich
STAT1 human, recombinant, expressed in insect cells, ≥70% (SDS-PAGE)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00