MilliporeSigma
  • 2-furoyl-LIGRLO-amide: a potent and selective proteinase-activated receptor 2 agonist.
2-furoyl-LIGRLO-amide: a potent and selective proteinase-activated receptor 2 agonist.
The Journal of pharmacology and experimental therapeutics (2004-02-21)
John J McGuire, Mahmoud Saifeddine, Chris R Triggle, Kimberly Sun, Morley D Hollenberg
ABSTRACT
A peptide corresponding to a proteinase-activated receptor 2 (PAR(2))-activating peptide with an N-terminal furoyl group modification, 2-furoyl-LIGRLO-NH(2), was assessed for PAR(2)-dependent and -independent biological activities. 2-Furoyl-LIGRLO-NH(2) was equally effective to and 10 to 25 times more potent than SLIGRLNH(2) for increasing intracellular calcium in cultured human and rat PAR(2)-expressing cells, respectively. In bioassays of tissue PAR(2) activity, measured as arterial vasodilation and hyperpolarization, 2-furoyl-LIGRLO-NH(2) was 10 to 300 times more potent than SLIGRL-NH(2). Unlike trans-cinnamoyl-LIGRLO-NH(2), 2-furoyl-LI-GRLO-NH(2) did not cause a prominent non-PAR(2)-mediated contraction of murine femoral arteries. In conclusion, 2-furoyl-LI-GRLO-NH(2) represents the most potent and selective activator of PAR(2) in biological systems described to date.
Sigma-Aldrich
2-Furoyl-LIGRLO-amide trifluoroacetate salt, ≥97% (HPLC)