MilliporeSigma
Novel ex-vivo mechanobiological intervertebral disc culture system.
Journal of biomechanics (2011-11-22)
Robert A Hartman, Kevin M Bell, Richard E Debski, James D Kang, Gwendolyn A Sowa
ABSTRACT
Intervertebral disc degeneration, a leading cause of low back pain, poses a significant socioeconomic burden with a broad array of costly treatment options. Mechanical loading is important in disease progression and treatment. Connecting mechanics and biology is critical for determining how loading parameters affect cellular response and matrix homeostasis. A novel ex-vivo experimental platform was developed to facilitate in-situ loading of rabbit functional spinal units (FSUs) with relevant biological outcome measures. The system was designed for motion outside of an incubator and validated for rigid fixation and physiologic environmental conditions. Specimen motion relative to novel fixtures was assessed using a digitizer; fixture stiffness exceeded specimen stiffness by an order of magnitude. Intradiscal pressure (IDP), measured using a fiber-optic pressure transducer, confirmed rigidity and compressive force selection. Surrounding media was controlled at 37 °C, 5% O(2)/CO(2) using a closed flow loop with an hypoxic incubator and was validated with probes in the specimen chamber. FSUs were subjected to cyclic compression (20 cycles) and four-hour creep at 1.0 MPa. Disc tissue was analyzed for cell viability using 3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT), which showed high viability (>90%) regardless of loading. Conditioned media was assayed for type-II collagen degradation fragments (CTX-II) and an aggrecan epitope (CS-846) associated with new aggrecan synthesis. CTX-II concentrations were not associated with loading, but CS-846 concentrations appeared to be increased with loading. Preservation of the full FSU allows physiologic load transmission and future multi-axis motion and identification of load-responsive proteins, thereby forming a new niche in intervertebral disc organ culture.
Sigma-Aldrich
2-Methoxyethanol, suitable for HPLC, ≥99.9%