MilliporeSigma
  • Dyskeratosis congenita: advances in the understanding of the telomerase defect and the role of stem cell transplantation.
Dyskeratosis congenita: advances in the understanding of the telomerase defect and the role of stem cell transplantation.
Pediatric transplantation (2007-08-01)
Josu de la Fuente, Inderjeet Dokal
ABSTRACT
DC is a multisystem bone marrow failure syndrome exhibiting marked clinical and genetic heterogeneity. X-linked, autosomal dominant and autosomal recessive subtypes are recognized. The gene mutated in X-linked DC (DKC1) encodes a highly conserved nucleolar protein called dyskerin. Dyskerin associates with the H/ACA motif class of small nucleolar RNAs in small nucleolar ribonucleoprotein particles that are important in guiding the conversion of uracil to pseudouracil during the maturation of ribosomal RNA. Dyskerin also associates with the TERC, which is important in the maintenance of telomeres. Mutations in TERC have been identified in patients with autosomal dominant DC and in a subset of patients with aplastic anemia and myelodysplasia. Recently, heterozygous mutations in TERT have been found in some patients with autosomal dominant DC and aplastic anemia. Additionally, patients with the severe multisystem disorder, Hoyeraal-Hreidarsson syndrome, have been found to have DKC1 mutations. Collectively, these observations have demonstrated that classical DC, Hoyeraal-Hreidarsson syndrome and a subset of aplastic anemia are due to a primary defect in telomerase. The critical role of telomeres and telomerase in humans is seen in the multisystem abnormalities found in these patients, including the increased incidence of malignancy. As bone marrow failure is the principal cause of death, conventional allografts have been attempted with limited success due to the high rate of pulmonary and endothelial complications. However, outcomes have improved with the use of non-myeloablative protocols, although the follow up is too short to evaluate long term toxicity and the natural course of the disease and it may be that correction of the telomerase defect is essential for the treatment of these patients.