MilliporeSigma
  • A purine nucleotide biosynthesis enzyme guanosine monophosphate reductase is a suppressor of melanoma invasion.
A purine nucleotide biosynthesis enzyme guanosine monophosphate reductase is a suppressor of melanoma invasion.
Cell reports (2013-10-22)
Joseph A Wawrzyniak, Anna Bianchi-Smiraglia, Wiam Bshara, Sudha Mannava, Jeff Ackroyd, Archis Bagati, Angela R Omilian, Michael Im, Natalia Fedtsova, Jeffrey C Miecznikowski, Kalyana C Moparthy, Shoshanna N Zucker, Qianqian Zhu, Nadezhda I Kozlova, Albert E Berman, Keith S Hoek, Andrei V Gudkov, Donna S Shewach, Carl D Morrison, Mikhail A Nikiforov
ABSTRACT
Melanoma is one of the most aggressive types of human cancers, and the mechanisms underlying melanoma invasive phenotype are not completely understood. Here, we report that expression of guanosine monophosphate reductase (GMPR), an enzyme involved in de novo biosynthesis of purine nucleotides, was downregulated in the invasive stages of human melanoma. Loss- and gain-of-function experiments revealed that GMPR downregulates the amounts of several GTP-bound (active) Rho-GTPases and suppresses the ability of melanoma cells to form invadopodia, degrade extracellular matrix, invade in vitro, and grow as tumor xenografts in vivo. Mechanistically, we demonstrated that GMPR partially depletes intracellular GTP pools. Pharmacological inhibition of de novo GTP biosynthesis suppressed whereas addition of exogenous guanosine increased invasion of melanoma cells as well as cells from other cancer types. Our data identify GMPR as a melanoma invasion suppressor and establish a link between guanosine metabolism and Rho-GTPase-dependent melanoma cell invasion.
Sigma-Aldrich
Anti-IMPDH2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GMPR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution