MilliporeSigma
  • Analysis of single nucleotide polymorphisms and haplotypes in HOXC gene cluster within susceptible region 12q13 of simple congenital heart disease.
Analysis of single nucleotide polymorphisms and haplotypes in HOXC gene cluster within susceptible region 12q13 of simple congenital heart disease.
Zhonghua yi xue yi chuan xue za zhi = Zhonghua yixue yichuanxue zazhi = Chinese journal of medical genetics (2005-10-11)
Li-guo Gong, Guang-rong Qiu, Hui Jiang, Xiao-yan Xu, Hong-yu Zhu, Kai-lai Sun
ABSTRACT
In the candidate region 12q13 of simple congenital heart disease(CHD), four single nucleotide polymorphisms(SNPs) in HOXC4 gene were chosen in order to investigate the distribution of SNP and haplotypes in simple CHD patients and normal people. The genotype of 4 SNPs in 108 simple CHD patients and 200 normal people were analyzed by restriction fragment length polymorphism(RFLP) and denaturing high-performance liquid chromatography(DHPLC). The statistical contingency table method was used to analyze SNP genotype frequency and gene frequency in patients and control group; then, the haplotypes were established and their frequencies in the two groups were assessed by PHASE software. C16476T polymorphism was not detected; A17860G located in 3' flanking sequence of HOXC5 gene displayed significant difference between the two groups. The G allele frequency in simple CHD patients was higher than that in healthy controls(P < 0.05); the distribution of frequencies of 4 haplotypes showed significant difference(P < 0.01). The A17860G located in 3'flanking sequence of HOXC5 gene is associated with simple CHD; the risk of CHD in the persons with G17860 is higher than that in those with A17860. the haplotype of 3 SNPs may be linked with the susceptible gene of simple CHD.