Merck
  • Lentivirus-Mediated knockdown of tectonic family member 1 inhibits medulloblastoma cell proliferation.
Lentivirus-Mediated knockdown of tectonic family member 1 inhibits medulloblastoma cell proliferation.
International journal of clinical and experimental medicine (2015-11-10)
Junjie Jing, Chengfeng Wang, Qinchuan Liang, Yang Zhao, Qingshuang Zhao, Shousen Wang, Jie Ma
ABSTRACT
Tectonic family member 1 (TCTN1) encodes a member of the tectonic family which are evolutionarily conserved secreted and transmembrane proteins, involving in a diverse variety of developmental processes. It has been demonstrated that tectonics expressed in regions that participate in Hedgehog (Hh) signaling during mouse embryonic development and was imperative for Hh-mediated patterning of the ventral neural tube. However, the expression and regulation of tectonics in human tumor is still not clear. In this study, shRNA-expressing lentivirus was constructed to knockdown TCTN1 in medulloblastoma cell line Daoy. The results showed that knockdown of TCTN1 inhibited cell proliferation and colony formation in Daoy cell line, also caused cell cycle arrest at the G2/M boundary. Taken all together, our data suggest that TCTN1 might play an important role in the progression of medulloblastoma.
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, non-essential amino acids and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture