MilliporeSigma
  • Gold nanoparticles-decorated graphene field-effect transistor biosensor for femtomolar MicroRNA detection.
Gold nanoparticles-decorated graphene field-effect transistor biosensor for femtomolar MicroRNA detection.
Biosensors & bioelectronics (2015-07-15)
Bingjie Cai, Le Huang, Hong Zhang, Zhongyue Sun, Zhiyong Zhang, Guo-Jun Zhang
ABSTRACT
Early detection is proven to be the best chance for successful cancer treatment. MiRNAs, as ideal biomarkers, can identify cancer in the early stage. Therefore, development of highly sensitive and selective detection methods for miRNA is still anticipated. Here we report on a gold nanoparticles (AuNPs)-decorated graphene field-effect transistor (FET) biosensor for highly sensitive, selective and label-free detection of miRNA. The AuNPs-decorated graphene FET biosensor was fabricated by drop-casting the reduced graphene oxide (R-GO) suspension onto the sensor surface, and subsequently decorating AuNPs onto the surface of R-GO. After peptide nucleic acid (PNA) probe was immobilized on the AuNPs surface, miRNA detection was carried out via PNA-miRNA hybridization. It was found that the developed FET biosensor was able to achieve a detection limit as low as 10 fM. In addition, the biosensor enabled an accurate distinction of complementary miRNA from one-base mismatched miRNA and noncomplementary miRNA. What's more, this highly sensitive and selective assay was also applied to the detection of miRNA in serum samples, making it a potential method for diagnosis of gene-related diseases.
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Ethanolamine, JIS special grade, ≥99.0%
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Sigma-Aldrich
Ethanolamine, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Cysteamine hydrochloride, BioXtra
Sigma-Aldrich
Cysteamine hydrochloride, ≥98% (titration)