MilliporeSigma
  • Comparative floral spur anatomy and nectar secretion in four representatives of Ranunculaceae.
Comparative floral spur anatomy and nectar secretion in four representatives of Ranunculaceae.
Protoplasma (2015-03-17)
Sebastian Antoń, Magdalena Kamińska
ABSTRACT
Nectaries are common in Ranunculaceae. These secretory structures, however, have not been studied in detail despite their importance in plant-animal interactions, and data relating to the structure of nectary spurs, which are so characteristic of several genera of this family, remain scarce. In order to redress this imbalance, we sought, in the present paper, to analyze the anatomical and ultrastructural organization of the nectary spurs of four representatives of Ranunculaceae, i.e., Aconitum lycoctonum L., Aquilegia vulgaris L., Consolida regalis Gray, and Delphinium elatum L. Nectary spurs were examined using light, fluorescence, scanning electron, and transmission electron microscopy. The floral nectaries of A. lycoctonum and A. vulgaris are situated at the apices of the spurs, whereas in C. regalis and D. elatum, the nectary is located along the floor surface of the spurs. Nectar in C. regalis and D. elatum is exuded through micro-channels in the cuticle, whereas in A. lycoctonum and A. vulgaris, it is released by means of cell wall disruption, indicating that the method of nectar secretion here is holocrine. Structurally, the nectary of all four investigated species is quite similar, and its cells are typical of nectar-producing cells described in the literature. It is proposed that in A. lycoctonum and A. vulgaris, disruption of the cell wall and the release of the entire cell contents into the spur cavity contribute to the composition of the nectar that the latter contains, enriching it with cytoplasmic components. We conclude that the manner of nectar exudation may vary considerably between closely related plant species, regardless of their geographical origin and phylogeny.
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, ≥99.5%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Pricing and Inventory is not available. Please check the Millipore website for more information.