MilliporeSigma
  • Compound mouse mutants of bZIP transcription factors Mafg and Mafk reveal a regulatory network of non-crystallin genes associated with cataract.
Compound mouse mutants of bZIP transcription factors Mafg and Mafk reveal a regulatory network of non-crystallin genes associated with cataract.
Human genetics (2015-04-22)
Smriti A Agrawal, Deepti Anand, Archana D Siddam, Atul Kakrana, Soma Dash, David A Scheiblin, Christine A Dang, Anne M Terrell, Stephanie M Waters, Abhyudai Singh, Hozumi Motohashi, Masayuki Yamamoto, Salil A Lachke
ABSTRACT
Although majority of the genes linked to early-onset cataract exhibit lens fiber cell-enriched expression, our understanding of gene regulation in these cells is limited to function of just eight transcription factors and largely in the context of crystallins. We report on small Maf transcription factors Mafg and Mafk as regulators of several non-crystallin human cataract-associated genes in fiber cells and establish their significance to this disease. We applied a bioinformatics tool for cataract gene discovery iSyTE to identify Mafg and its co-regulators in the lens, and generated various null-allelic combinations of Mafg:Mafk mouse mutants for phenotypic and molecular analysis. By age 4 months, Mafg-/-:Mafk+/- mutants exhibit lens defects that progressively develop into cataract. High-resolution phenotypic characterization of Mafg-/-:Mafk+/- mouse lens reveals severely disorganized fiber cells, while microarray-based expression profiling identifies 97 differentially regulated genes (DRGs). Integrative analysis of Mafg-/-:Mafk+/- lens-DRGs with (1) binding motifs and genomic targets of small Mafs and their regulatory partners, (2) iSyTE lens expression data, and (3) interactions between DRGs in the String database, unravel a detailed small Maf regulatory network in the lens, several nodes of which are linked to cataract. This approach identifies 36 high-priority candidates from the original 97 DRGs. Significantly, 8/36 (22%) DRGs are associated with cataracts in human (GSTO1, MGST1, SC4MOL, UCHL1) or mouse (Aldh3a1, Crygf, Hspb1, Pcbd1), suggesting a multifactorial etiology that includes oxidative stress and misregulation of sterol synthesis. These data identify Mafg and Mafk as new cataract-associated candidates and define their function in regulating largely non-crystallin genes linked to human cataract.
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Carbon, mesoporous
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.9 atom % 12C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Carbon nanofibers, graphitized, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Sigma-Aldrich
Carbon nanofibers, pyrolitically stripped, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
Estimated to ship on March 24, 2025
$518.00