MilliporeSigma
  • Fitting accuracy and fracture resistance of crowns using a hybrid zirconia frame made of both porous and dense zirconia.
Fitting accuracy and fracture resistance of crowns using a hybrid zirconia frame made of both porous and dense zirconia.
Dental materials journal (2015-03-06)
Takashi Nakamura, Tsuyoshi Sugano, Hirofumi Usami, Kazumichi Wakabayashi, Hiroshi Ohnishi, Tohru Sekino, Hirofumi Yatani
ABSTRACT
The purpose of this study is to evaluate the fitting accuracy and fracture resistance of crowns using a hybrid zirconia frame made of both porous and dense zirconia. Commercial semi-sintered zirconia, sintered dense zirconia and sintered hybrid zirconia were used. Sintered zirconia was milled using the CAD/CAM system, and semi-sintered zirconia was milled and sintered to fabricate molar crown frames. Completed frames were veneered with tooth-colored porcelain. The marginal and internal gaps between frames/crowns and abutments were measured. Each crown specimen was subjected to a fracture test. There were no significant differences in marginal and internal gap among all the frames and crowns. The crown with the hybrid zirconia frame had a 31-35% greater fracture load than that with the commercial or dense zirconia frame (p<0.01). This suggests that the all-ceramic crowns with a hybrid zirconia frame have a high fracture resistance.
Sigma-Aldrich
Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00