Merck
  • Subcritical extraction of flaxseed oil with n-propane: Composition and purity.
Subcritical extraction of flaxseed oil with n-propane: Composition and purity.
Food chemistry (2015-06-05)
Ana Beatriz Zanqui, Damila Rodrigues de Morais, Claudia Marques da Silva, Jandyson Machado Santos, Sandra Terezinha Marques Gomes, Jesuí Vergílio Visentainer, Marcos Nogueira Eberlin, Lúcio Cardozo-Filho, Makoto Matsushita
ABSTRACT
Flaxseed (Linum usitatissimum L.) oil was obtained via subcritical n-propane fluid extraction (SubFE) under different temperatures and pressures with an average yield of 28% and its composition, purity and oxidative stability were compared to oils obtained via conventional solvent extraction methods (SEMs). When the oxidative stability was measured by differential scanning calorimetry, the oil was found to be up to 5 times more resistant to lipid oxidation as compared to the SEM oils. Direct infusion electrospray ionization mass spectrometry (ESI-MS) analysis showed characteristic and similar TAG profiles for SubFE and SEMs oils but higher purity for the SubFE oil. The flaxseed oil content of β-tocopherol, campesterol, stigmasterol and sitosterol were quantified via GC-MS. SubFE showed to be a promising alternative to conventional SEM since SubFE provides an oil with higher purity and higher oxidation stability and with comparable levels of biologically active components.
Sigma-Aldrich
Methanol, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl tricosanoate, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5-α-Cholestane, ≥97.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, LiChropur
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propane, 99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propane, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trifluoroacetamide, 97%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%